MilliporeSigma
  • A histological study of the song system of the carrion crow (Corvus corone).
A histological study of the song system of the carrion crow (Corvus corone).
The Journal of comparative neurology (2021-01-22)
Ylva Kersten, Bettina Friedrich-Müller, Andreas Nieder
ABSTRACT
The song system of songbirds (oscines) is one of the best studied neuroethological model systems. So far, it has been treated as a relatively constrained sensorimotor system. Songbirds such as crows, however, are also known for their capability to cognitively control their audio-vocal system. Yet, the neuroanatomy of the corvid song system has never been explored systematically. We aim to close this scientific gap by presenting a stereotactic investigation of the extended song system of the carrion crow (Corvus corone), an oscine songbird of the corvid family that has become an interesting model system for cognitive neuroscience. In order to identify and delineate the song nuclei, the ascending auditory nuclei, and the descending vocal-motor nuclei, four stains were applied. In addition to the classical Nissl-, myelin-, and a combination of Nissl-and-myelin staining, staining for tyrosine hydroxylase was used to reveal the distribution of catecholaminergic neurons (dopaminergic, noradrenergic, and adrenergic) in the song system. We show that the crow brain contains the important song-related nuclei, including auditory input and motor output structures, and map them throughout the brain. Fiber-stained sections reveal putative connection patterns between the crow's song nuclei comparable to other songbirds.
Sigma-Aldrich
Anti-Mouse IgG (H+L), highly cross adsorbed-Biotin antibody produced in goat, affinity isolated antibody, lyophilized powder