MilliporeSigma
  • Reversal of tumoral immune resistance by inhibition of tryptophan 2,3-dioxygenase.
Reversal of tumoral immune resistance by inhibition of tryptophan 2,3-dioxygenase.
Proceedings of the National Academy of Sciences of the United States of America (2012-02-07)
Luc Pilotte, Pierre Larrieu, Vincent Stroobant, Didier Colau, Eduard Dolusic, Raphaël Frédérick, Etienne De Plaen, Catherine Uyttenhove, Johan Wouters, Bernard Masereel, Benoît J Van den Eynde
ABSTRACT
Tryptophan catabolism mediated by indoleamine 2,3-dioxygenase (IDO1) is an important mechanism of peripheral immune tolerance contributing to tumoral immune resistance, and IDO1 inhibition is an active area of drug development. Tryptophan 2,3-dioxygenase (TDO) is an unrelated hepatic enzyme that also degrades tryptophan along the kynurenine pathway. Here, we show that enzymatically active TDO is expressed in a significant proportion of human tumors. In a preclinical model, TDO expression by tumors prevented their rejection by immunized mice. We developed a TDO inhibitor, which, upon systemic treatment, restored the ability of mice to reject TDO-expressing tumors. Our results describe a mechanism of tumoral immune resistance based on TDO expression and establish proof-of-concept for the use of TDO inhibitors in cancer therapy.
Sigma-Aldrich
LM10, ≥98% (HPLC)