MilliporeSigma
  • Transcriptional Repression of Aerobic Glycolysis by OVOL2 in Breast Cancer.
Transcriptional Repression of Aerobic Glycolysis by OVOL2 in Breast Cancer.
Advanced science (Weinheim, Baden-Wurttemberg, Germany) (2022-07-28)
Xiujuan Zhang, Fei Luo, Shaliu Luo, Ling Li, Xinxin Ren, Jing Lin, Yingchun Liang, Chao Ma, Lihua Ding, Deyu Zhang, Tianxing Ye, Yanni Lin, Bilian Jin, Shan Gao, Qinong Ye
ABSTRACT
Aerobic glycolysis (Warburg effect), a hallmark of cancer, plays a critical role in cancer cell growth and metastasis; however, direct inhibition of the Warburg effect remains largely unknown. Herein, the transcription factor OVO-like zinc finger 2 (OVOL2) is demonstrated to directly repress the expression of several glycolytic genes, blocking the Warburg effect and breast tumor growth and metastasis in vitro and in vivo. OVOL2 inhibits glycolysis by recruiting the nuclear receptor co-repressor (NCoR) and histone deacetylase 3 (HDAC3). The tumor suppressor p53, a key regulator of cancer metabolism, activates OVOL2 by binding to the oncoprotein mouse double minute 2 homolog (MDM2) and inhibiting MDM2-mediated ubiquitination and degradation of OVOL2. OVOL2 expression is negatively correlated with glycolytic gene expression and can be a good predictor of prognosis in patients with breast cancer. Therefore, targeting the p53/MDM2/OVOL2 axis provides a potential avenue for cancer treatment, especially breast cancer.
Sigma-Aldrich
Monoclonal Anti-GAPDH−Peroxidase antibody produced in mouse, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Millipore
EZview Red Anti-c-Myc Affinity Gel
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys4) - ChIP Validated Antibody and Primer Set, from rabbit, purified by affinity chromatography
Sigma-Aldrich
ChIPAb+ N-CoR Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-c-Myc−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
ChIPAb+ HDAC1 - ChIP Validated Antibody and Primer Set, culture supernatant, from mouse
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 (Lys27) - ChIP Validated Antibody and Primer Set, clone CMA309, from mouse
Sigma-Aldrich
ChIPAb+ p53 - ChIP Validated Antibody and Primer Set, from mouse