MilliporeSigma
  • GUB06-046, a novel secretin/glucagon-like peptide 1 co-agonist, decreases food intake, improves glycemic control, and preserves beta cell mass in diabetic mice.
GUB06-046, a novel secretin/glucagon-like peptide 1 co-agonist, decreases food intake, improves glycemic control, and preserves beta cell mass in diabetic mice.
Journal of peptide science : an official publication of the European Peptide Society (2017-10-24)
Søren B van Witteloostuijn, Louise S Dalbøge, Gitte Hansen, Søren Roi Midtgaard, Grethe Vestergaard Jensen, Knud J Jensen, Niels Vrang, Jacob Jelsing, Søren L Pedersen
ABSTRACT
Bariatric surgery is currently the most effective treatment of obesity, which has spurred an interest in developing pharmaceutical mimetics. It is thought that the marked body weight-lowering effects of bariatric surgery involve stimulated secretion of appetite-regulating gut hormones, including glucagon-like peptide 1. We here report that intestinal expression of secretin is markedly upregulated in a rat model of Roux-en-Y gastric bypass, suggesting an additional role of secretin in the beneficial metabolic effects of Roux-en-Y gastric bypass. We therefore developed novel secretin-based peptide co-agonists and identified a lead compound, GUB06-046, that exhibited potent agonism of both the secretin receptor and glucagon-like peptide 1 receptor. Semi-acute administration of GUB06-046 to lean mice significantly decreased cumulative food intake and improved glucose tolerance. Chronic administration of GUB06-046 to diabetic db/db mice for 8 weeks improved glycemic control, as indicated by a 39% decrease in fasting blood glucose and 1.6% reduction of plasma HbA1c levels. Stereological analysis of db/db mice pancreata revealed a 78% increase in beta-cell mass after GUB06-046 treatment, with no impact on exocrine pancreas mass or pancreatic duct epithelial mass. The data demonstrate beneficial effects of GUB06-046 on appetite regulation, glucose homeostasis, and beta-cell mass in db/db mice, without proliferative effects on the exocrine pancreas and the pancreatic duct epithelium. Copyright © 2017 European Peptide Society and John Wiley & Sons, Ltd.
Sigma-Aldrich
MMP Inhibitor V, The MMP Inhibitor V, also referenced under CAS 223472-31-9, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
MMP Inhibitor III, The MMP Inhibitor III, also referenced under CAS 927827-98-3, controls the biological activity of MMP. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.