MilliporeSigma
  • Systematic Exploration of Passive Permeability in Tetrapeptides with Hydrogen-Bond-Accepting Amino Acid Side Chains.
Systematic Exploration of Passive Permeability in Tetrapeptides with Hydrogen-Bond-Accepting Amino Acid Side Chains.
ChemMedChem (2022-06-14)
Hiroki Shimizu, Adam R Renslo
ABSTRACT
We synthesized and experimentally tested the passive permeability of more than thirty tetrapeptides mimicking the N-terminus of the pro-apoptotic protein Smac (Second mitochondria-derived activator of caspases). Each peptide bore one or two unnatural Hydrogen Bond Acceptor-bearing Amino Acid (HBA-AA) residues, such that intramolecular hydrogen bonding with proximal backbone amide N-H donors is feasible. Passive permeability of the synthetic peptides was determined using the parallel artificial membrane permeability assay (PAMPA). Experimental permeability values were found to span three orders of magnitude, providing useful empirical guidance for the design of more permeable Smac mimetics specifically, and peptidic ligands generally.
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Cimetidine
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder