MilliporeSigma
  • The tryptophan metabolizing enzyme indoleamine 2,3-dioxygenase 1 regulates polycystic kidney disease progression.
The tryptophan metabolizing enzyme indoleamine 2,3-dioxygenase 1 regulates polycystic kidney disease progression.
JCI insight (2022-11-25)
Dustin T Nguyen, Emily K Kleczko, Nidhi Dwivedi, Marie-Louise T Monaghan, Berenice Y Gitomer, Michel B Chonchol, Eric T Clambey, Raphael A Nemenoff, Jelena Klawitter, Katharina Hopp
ABSTRACT
Autosomal dominant polycystic kidney disease (ADPKD), the most common monogenic nephropathy, is characterized by phenotypic variability exceeding genic effects. Dysregulated metabolism and immune cell function are key disease modifiers. The tryptophan metabolites, kynurenines, produced through IDO1, are known immunomodulators. Here, we study the role of tryptophan metabolism in PKD using an orthologous disease model (C57Bl/6J Pkd1RC/RC). We found elevated kynurenine and IDO1 levels in Pkd1RC/RC kidneys versus wildtype. Further, IDO1 levels were increased in ADPKD cell lines. Genetic Ido1 loss in Pkd1RC/RC animals resulted in reduced PKD severity as measured by %kidney weight/body weight and cystic index. Consistent with an immunomodulatory role of kynurenines, Pkd1RC/RC;Ido1-/- mice presented with significant changes in the cystic immune microenvironment (CME) versus controls. Kidney macrophage numbers decreased and CD8+ T cell numbers increased, both known PKD modulators. Also, pharmacological IDO1 inhibition in Pkd1RC/RC mice and kidney specific Pkd2 knockout mice with rapidly progressive PKD resulted in less severe PKD versus controls with similar changes in the CME as in the genetic model. Our data suggest that tryptophan metabolism is dysregulated in ADPKD and that its inhibition results in changes to the CME and slows disease progression, making IDO1 a novel therapeutic target for ADPKD.
Sigma-Aldrich
1-Methyl-D-tryptophan, 95%
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution