MilliporeSigma
  • Covalent perturbation as a tool for validation of identifications and PTM mapping applied to bovine alpha-crystallin.
Covalent perturbation as a tool for validation of identifications and PTM mapping applied to bovine alpha-crystallin.
Proteomics (2015-12-09)
Jakob Bunkenborg, Lasse Gaarde Falkenby, Lea Mørch Harder, Henrik Molina
ABSTRACT
Proteomic identifications hinge on the measurement of both parent and fragment masses and matching these to amino acid sequences via database search engines. The correctness of the identifications is assessed by statistical means. Here we present an experimental approach to test identifications. Chemical modification of all peptides in a sample leads to shifts in masses depending on the chemical properties of each peptide. The identification of a native peptide sequence and its perturbed version with a different parent mass and fragment ion masses provides valuable information. Labeling all peptides using reductive alkylation with formaldehyde is one such perturbation where the ensemble of peptides shifts mass depending on the number of reactive amine groups. Matching covalently perturbed fragmentation patterns from the same underlying peptide sequence increases confidence in the assignments and can salvage low scoring post-translationally modified peptides. Applying this strategy to bovine alpha-crystallin, we identify 9 lysine acetylation sites, 4 O-GlcNAc sites and 13 phosphorylation sites.