MilliporeSigma
  • Ketogenic HMG-CoA lyase and its product β-hydroxybutyrate promote pancreatic cancer progression.
Ketogenic HMG-CoA lyase and its product β-hydroxybutyrate promote pancreatic cancer progression.
The EMBO journal (2022-03-22)
Victoire Gouirand, Tristan Gicquel, Evan C Lien, Emilie Jaune-Pons, Quentin Da Costa, Pascal Finetti, Elodie Metay, Camille Duluc, Jared R Mayers, Stephane Audebert, Luc Camoin, Laurence Borge, Marion Rubis, Julie Leca, Jeremy Nigri, François Bertucci, Nelson Dusetti, Juan L Iovanna, Richard Tomasini, Ghislain Bidaut, Fabienne Guillaumond, Matthew G Vander Heiden, Sophie Vasseur
ABSTRACT
Pancreatic ductal adenocarcinoma (PDA) tumor cells are deprived of oxygen and nutrients and therefore must adapt their metabolism to ensure proliferation. In some physiological states, cells rely on ketone bodies to satisfy their metabolic needs, especially during nutrient stress. Here, we show that PDA cells can activate ketone body metabolism and that β-hydroxybutyrate (βOHB) is an alternative cell-intrinsic or systemic fuel that can promote PDA growth and progression. PDA cells activate enzymes required for ketogenesis, utilizing various nutrients as carbon sources for ketone body formation. By assessing metabolic gene expression from spontaneously arising PDA tumors in mice, we find HMG-CoA lyase (HMGCL), involved in ketogenesis, to be among the most deregulated metabolic enzymes in PDA compared to normal pancreas. In vitro depletion of HMGCL impedes migration, tumor cell invasiveness, and anchorage-independent tumor sphere compaction. Moreover, disrupting HMGCL drastically decreases PDA tumor growth in vivo, while βOHB stimulates metastatic dissemination to the liver. These findings suggest that βOHB increases PDA aggressiveness and identify HMGCL and ketogenesis as metabolic targets for limiting PDA progression.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Anti-BDH2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Roche
Mitomycin C, crystalline powder, 98%, pkg of 2 mg
Sigma-Aldrich
Anti-ACAT1 antibody produced in rabbit, affinity isolated antibody