MilliporeSigma
  • Marked disruption and aberrant regulation of elastic fibres in early striae gravidarum.
Marked disruption and aberrant regulation of elastic fibres in early striae gravidarum.
The British journal of dermatology (2015-07-17)
F Wang, K Calderone, N R Smith, T T Do, Y R Helfrich, T R B Johnson, S Kang, J J Voorhees, G J Fisher
ABSTRACT
Striae gravidarum (SG), or 'stretch marks' of pregnancy, begin as erythematous streaks, and mature over months to years to become permanent scar-like bands that may be hypopigmented, atrophic and lax. To investigate early molecular alterations that may promote laxity of mature SG, we investigated the dermal elastic fibre network, which provides human skin with elastic properties. We obtained skin samples of newly developed, erythematous abdominal SG in healthy pregnant women. The elastic fibre network was examined by Verhoeff elastic staining and immunofluorescence staining of skin sections. Gene expression was measured by real-time polymerase chain reaction. The normal elastic fibre network appeared markedly disrupted in SG, compared with perilesional abdominal skin or control (normal-appearing hip skin). This disruption was accompanied by the emergence of short, disorganized, thin, thread-like 'fibrils', which were observed prominently in the mid-to-deep dermis. These fibrils were rich in tropoelastin (the main component of normal elastic fibres), and persisted into the postpartum period without forming normal-appearing elastic fibres. The emergence of these fibrils was accompanied by increased gene expression of tropoelastin and fibrillin-1, but not other elastic fibre components, including fibrillin-2 and fibulin-1, -2 or -5. In early SG, the elastic fibre network appears markedly disrupted, and newly synthesized tropoelastin-rich fibrils emerge, likely as a result of uncoordinated synthesis of elastic fibre components. Because they are thin and disorganized, tropoelastin-rich fibrils likely do not function as normal elastic fibres do. These observations provide the foundations for elucidating pathogenic mechanisms by which laxity may develop in SG.
Sigma-Aldrich
Anti-Fibrillin Antibody, clone 11C1.3, clone 11C1.3, Chemicon®, from mouse