MilliporeSigma
  • Protection by -Biotics against Hypertension Programmed by Maternal High Fructose Diet: Rectification of Dysregulated Expression of Short-Chain Fatty Acid Receptors in the Hypothalamic Paraventricular Nucleus of Adult Offspring.
Protection by -Biotics against Hypertension Programmed by Maternal High Fructose Diet: Rectification of Dysregulated Expression of Short-Chain Fatty Acid Receptors in the Hypothalamic Paraventricular Nucleus of Adult Offspring.
Nutrients (2022-10-28)
Yung-Mei Chao, You-Lin Tain, Wei-Chia Lee, Kay L H Wu, Hong-Ren Yu, Julie Y H Chan
ABSTRACT
The role of short-chain fatty acids (SCFAs) in the brain on the developmental programming of hypertension is poorly understood. The present study explored dysregulated tissue levels of SCFAs and expression of SCFA-sensing receptors in the hypothalamic paraventricular nucleus (PVN), a key forebrain region engaged in neural regulation of blood pressure of offspring to maternal high fructose diet (HFD) exposure. We further investigated the engagement of SCFA-sensing receptors in PVN in the beneficial effects of -biotics (prebiotic, probiotic, synbiotic, and postbiotic) on programmed hypertension. Maternal HFD during gestation and lactation significantly reduced circulating butyrate, along with decreased tissue level of butyrate and increased expression of SCFA-sensing receptors, GPR41 and olfr78, and tissue oxidative stress and neuroinflammation in PVN of HFD offspring that were rectified by oral supplement with -biotics. Gene silencing of GPR41 or olfr78 mRNA in PVN also protected adult HFD offspring from programmed hypertension and alleviated the induced oxidative stress and inflammation in PVN. In addition, oral supplement with postbiotic butyrate restored tissue butyrate levels, rectified expressions of GPR41 and olfr78 in PVN, and protected against programmed hypertension in adult HFD offspring. These data suggest that alterations in tissue butyrate level, expression of GPR41 and olfr78, and activation of SCFA-sensing receptor-dependent tissue oxidative stress and neuroinflammation in PVN could be novel mechanisms that underlie hypertension programmed by maternal HFD exposure in adult offspring. Furthermore, oral -biotics supplementation may exert beneficial effects on hypertension of developmental origin by targeting dysfunctional SCFA-sensing receptors in PVN to exert antioxidant and anti-inflammatory actions in the brain.
Sigma-Aldrich
Anti-FFAR2 Antibody/GPR43, from rabbit, purified by affinity chromatography