MilliporeSigma
  • Allotype analysis to determine the origin of cytomegalovirus immunoglobulin-G after allogeneic stem cell transplantation.
Allotype analysis to determine the origin of cytomegalovirus immunoglobulin-G after allogeneic stem cell transplantation.
Transplant infectious disease : an official journal of the Transplantation Society (2014-11-05)
R Yamazaki, Y Tanaka, H Nakasone, M Sato, K Terasako-Saito, K Sakamoto, Y Akahoshi, H Nakano, T Ugai, R Yamasaki, H Wada, Y Ishihara, K Kawamura, M Ashizawa, S-I Kimura, M Kikuchi, S Kako, J Kanda, A Tanihara, J Nishida, Y Kanda
ABSTRACT
Cytomegalovirus (CMV) reactivation still remains a major problem following allogeneic hematopoietic stem cell transplantation (HSCT). In this study, we analyzed an immunoglobulin allotype, IgG1m(f), in CMV-seropositive HSCT recipients and their donors to distinguish donor-derived antibody from recipient-derived antibody. Eight donor-recipient pairs were informative regarding the appearance of donor-derived immunoglobulin-G (IgG), as the recipients were homozygous null for the IgG1m(f) allotype and the donors were IgG1m(f) positive. In these patients, total IgG, IgM, and allotype-specific IgG against CMV were measured by enzyme-linked immunosorbent assay. All subjects were monitored for at least 9 months after HSCT with (n = 5) or without (n = 3) CMV reactivation. Donor-derived CMV IgG tended to be elevated earlier in patients with CMV-seropositive donors than in those with CMV-seronegative donors. In 1 patient with a CMV-negative donor, donor-derived CMV IgG was not detected until late CMV reactivation. In 3 patients without CMV reactivation, donor-derived CMV IgG was also elevated within 1-6 months after HSCT. In conclusion, the CMV serostatus of the donor may be related to the timing of the appearance of donor-derived CMV IgG and the reconstitution of humoral immunity against CMV, regardless of the CMV antigenemia level after HSCT.
Sigma-Aldrich
Monoclonal Anti-Human IgG1 (Fab specific) [G1m(f)] antibody produced in mouse, clone SG-16, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Human IgG (whole molecule)−Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution