MilliporeSigma
  • TET2 is required for Type I IFN-mediated inhibition of bat-origin swine acute diarrhea syndrome coronavirus.
TET2 is required for Type I IFN-mediated inhibition of bat-origin swine acute diarrhea syndrome coronavirus.
Journal of medical virology (2022-02-26)
Yueyue Duan, Cong Yuan, Xuepeng Suo, Liyan Cao, Xiangyu Kong, Xiangtong Li, Haixue Zheng, Qi Wang
ABSTRACT
Swine acute diarrhea syndrome coronavirus (SADS-CoV) is a newly discovered bat-origin coronavirus with fatal pathogenicity for neonatal piglets. There is no vaccine to prevent SADS-CoV infection or clinically approved drugs targeting SADS-CoV. Therefore, unraveling cellular factors that regulate SADS-CoV for cell entry is critical to understanding the viral transmission mechanism and provides a potential therapeutic target for SADS-CoV cure. Here, we showed that Type I interferon (IFN-I) pretreatment potently blocks SADS-CoV entry into cells using lentiviral pseudo-virions as targets whose entry is driven by the SADS-CoV Spike glycoprotein. IFN-I-mediated inhibition of SADS-CoV entry and replication was dramatically impaired in the absence of TET2. These results suggest TET2 is found to serve as a checkpoint of IFN-I-meditated inhibition on the cell entry of SADS-CoV, and our discovery might constitute a novel treatment option to combat against SADS-CoV.
Sigma-Aldrich
Anti-TET2 Antibody, clone hT2H 21F11, clone hT2H21F11, from mouse
Pricing and availability is not currently available.