MilliporeSigma
  • Reproducing the scalp microbiota community: co-colonization of a 3D reconstructed human epidermis with C. acnes and M. restricta.
Reproducing the scalp microbiota community: co-colonization of a 3D reconstructed human epidermis with C. acnes and M. restricta.
International journal of cosmetic science (2021-01-21)
Marisa Meloni, Silvia Balzaretti, Nathalie Collard, Stephane Desaint, Celine Laperdrix
ABSTRACT
A 3D reconstructed human epidermis (RHE) model colonized with specific microbial strains was developed to model the complex interactions between strains of the human scalp hair. Reconstructed human epidermis was colonized with Cutibacterium acnes and Malassezia restricta for 72 h. The epidermal model was characterized in terms of morphology, using immune-labelling targeting biomarkers for barrier structure, proliferation, differentiation and anti-microbial defence. The barrier function was assessed by transepithelial electrical eesistance (TEER) measurements. In order to study the microorganisms on the epidermal model, viable counts and phenotype ultrastructure analysis were performed by scanning electron microscopy (SEM). The RHE colonized with C. acnes did not lead to severe modifications of the physiological barrier integrity and viability, though it shows aggregates. M. restricta formed large aggregates by a close interaction with the RHE, thus causing both a strong decrease in barrier function and structure degradation and an increased human beta defensin 2 (HBD2) expression. The co-colonized model resulted in barrier depletion, but the overall damage was less severe, respecting the single colonization with M. restricta. The developed 'scalp model' allowed to identify morphological modifications leading to uncontrolled epidermal renewal. This study shows a pre-clinical model that recapitulates the interactions that can occur between site-specific microbial strains and keratinocytes in dandruff condition. The model can be applied to assess ingredients and products' mechanism of action.
Sigma-Aldrich
Anti-KRT10 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-IVL antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-KRT14 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution