MilliporeSigma
  • Interleukin-8 enhances myocilin expression, Akt-FoxO3 signaling and myogenic differentiation in rat skeletal muscle cells.
Interleukin-8 enhances myocilin expression, Akt-FoxO3 signaling and myogenic differentiation in rat skeletal muscle cells.
Journal of cellular physiology (2019-04-05)
Marta Milewska, Tomasz Domoradzki, Alicja Majewska, Maciej Błaszczyk, Małgorzata Gajewska, Magdalena Hulanicka, Anna Ciecierska, Katarzyna Grzelkowska-Kowalczyk
ABSTRACT
Interleukin (IL)-8 is released both in visceral adipose tissue and in contracting skeletal muscles. In this study, we examined cellular pathways associated with muscle hypertrophy, chosen on the basis of microRNA profiling, in differentiating rat primary skeletal muscle cells (RSkMC) treated with IL-8 (1 ng/ml) for 11 days. IL-8 increased myocilin expression, Akt phosphorylation, FoxO3 dispersion throughout the cytoplasm, and reduced FoxO3 level. IL-8 decreased the expression of atrogin and MuRF1 and increased myotube length and diameter. We concluded that IL-8 present in extracellular environment of myoblasts induced to differentiation stimulates expression of myocilin, a protein important for skeletal muscle hypertrophy. This phenomenon was associated with: (a) activation of myogenic transcription, (b) increased phosphorylation and activation of PKB/Akt, leading to (c) cytoplasm distribution and degradation of a transcription factor FoxO3, (d) decreased expression of gene markers of proteolysis, atrogin and Murf1, and (e) increased myotube length and diameter. In this regard, IL-8 affects skeletal muscle cells similarly to IGF-I and can be considered as a potent anticatabolic factor for skeletal muscle.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse