MilliporeSigma
  • Cell shape-based chemical screening reveals an epigenetic network mediated by focal adhesions.
Cell shape-based chemical screening reveals an epigenetic network mediated by focal adhesions.
The FEBS journal (2021-03-27)
Tomomi Kanazawa, Hiroki Michida, Yuki Uchino, Akari Ishihara, Suxiang Zhang, Sho Tabata, Yutaka Suzuki, Akira Imamoto, Mariko Okada
ABSTRACT
Adapter proteins CRK and CRKL participate in a variety of signaling pathways, including cell adhesion, and fate regulation of mammalian cells. However, the molecular functions of CRK/CRKL in epigenetic regulation remain largely unknown. Here, we developed a pipeline to evaluate cell morphology using high-content image analysis combined with chemical screening of kinase and epigenetic modulators. We found that CRK/CRKL modulates gene regulatory networks associated with cell morphology through epigenetic alteration in mouse embryonic fibroblasts. Integrated epigenome and transcriptome analyses revealed that CRK/CRKL is involved in super-enhancer activity and upregulation of Cdt1, Rin1, and Spp1 expression for the regulation of cell morphology. Screening of a library of 80 epigenetic inhibitors showed that histone H3 modifiers, euchromatic histone methyltransferase 2 and mitogen- and stress-activated kinase 1, may be important for CRK/CRKL-mediated morphological changes. Taken together, our results indicate that CRK/CRKL plays a critical role in gene regulatory networks through epigenetic modification. DATABASES: Chromatin immunoprecipitation sequencing and RNA sequencing data were deposited in the DNA Data Bank of Japan under DRA011080 and DRA011081 accession numbers, respectively.
Sigma-Aldrich
Anti-CrkL Antibody, clone 5-6, clone 5-6, Upstate®, from mouse