MilliporeSigma
  • Profilin-1 regulates DNA replication forks in a context-dependent fashion by interacting with SNF2H and BOD1L.
Profilin-1 regulates DNA replication forks in a context-dependent fashion by interacting with SNF2H and BOD1L.
Nature communications (2022-11-03)
Cuige Zhu, Mari Iwase, Ziqian Li, Faliang Wang, Annabel Quinet, Alessandro Vindigni, Jieya Shao
ABSTRACT
DNA replication forks are tightly controlled by a large protein network consisting of well-known core regulators and many accessory factors which remain functionally undefined. In this study, we report previously unknown nuclear functions of the actin-binding factor profilin-1 (PFN1) in DNA replication, which occur in a context-dependent fashion and require its binding to poly-L-proline (PLP)-containing proteins instead of actin. In unperturbed cells, PFN1 increases DNA replication initiation and accelerates fork progression by binding and stimulating the PLP-containing nucleosome remodeler SNF2H. Under replication stress, PFN1/SNF2H increases fork stalling and functionally collaborates with fork reversal enzymes to enable the over-resection of unprotected forks. In addition, PFN1 binds and functionally attenuates the PLP-containing fork protector BODL1 to increase the resection of a subset of stressed forks. Accordingly, raising nuclear PFN1 level decreases genome stability and cell survival during replication stress. Thus, PFN1 is a multi-functional regulator of DNA replication with exploitable anticancer potential.
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-SMARCA5 Antibody, from rabbit, purified by affinity chromatography
Millipore
Benzonase® Nuclease, Purity > 90%
Sigma-Aldrich
Anti-Rad51 (Ab-1) Rabbit pAb, liquid, Calbiochem®