MilliporeSigma
  • Deviant circadian rhythmicity, corticosterone variability and trait testosterone levels in aggressive mice.
Deviant circadian rhythmicity, corticosterone variability and trait testosterone levels in aggressive mice.
The European journal of neuroscience (2022-03-02)
Floriana Mogavero, Kitty van Zwieten, Jan K Buitelaar, Jeffrey C Glennon, Marloes J A G Henckens
ABSTRACT
Although aggression has been linked to disturbances of circadian rhythm, insight into the neural substrate of this association is currently lacking. The suprachiasmatic nucleus (SCN) of the hypothalamus, the master circadian clock, is regulated by clock genes and known to influence the secretion of cortisosterone and testosterone, important hormones implicated in aggression. Here, we investigated deviations in the regulation of the locomotor circadian rhythm and hormonal levels in a mouse model of abnormal aggression. We tested aggressive BALB/cJ and control BALB/cByJ mice in the resident-intruder paradigm and compared them on their locomotor circadian rhythm during a 12 h light/12 h dark cycle and constant darkness. State (serum) corticosterone and trait (hair) corticosterone and testosterone levels were determined, and immunohistochemistry was performed to assess the expression of important clock proteins, PER1 and PER2, in the core and shell of the SCN at the start of their active phase. Compared with BALB/cByJ mice, aggressive BALB/cJ mice displayed: (1) a shorter free-running period in constant darkness; (2) reduced state corticosterone variability between circadian peak and trough but no differences in corticosterone trait levels; (3) lower testosterone trait levels; (4) higher PER1 expression in the SCN shell with no changes in PER2 in either SCN subregion during the early dark phase. Together, these results suggest that aggressive BALB/cJ mice have disturbances in different components encompassing the circadian and hormonal cycle, emphasizing their value for future investigation of the causal relationship between SCN function, circadian clocks and aggression.
Sigma-Aldrich
Anti-mPER1 (residues 6-21) Antibody, serum, from rabbit