MilliporeSigma
  • An order of magnitude faster DNA-PAINT imaging by optimized sequence design and buffer conditions.
An order of magnitude faster DNA-PAINT imaging by optimized sequence design and buffer conditions.
Nature methods (2019-10-09)
Florian Schueder, Johannes Stein, Florian Stehr, Alexander Auer, Bianca Sperl, Maximilian T Strauss, Petra Schwille, Ralf Jungmann
ABSTRACT
DNA points accumulation in nanoscale topography (DNA-PAINT) is a relatively easy-to-implement super-resolution technique. However, image acquisition is slow compared to most other approaches. Here, we overcome this limitation by designing optimized DNA sequences and buffer conditions. We demonstrate our approach in vitro with DNA origami and in situ using cell samples, and achieve an order of magnitude faster imaging speeds without compromising image quality or spatial resolution. This improvement now makes DNA-PAINT applicable to high-throughput studies.
Sigma-Aldrich
Anti-GOLGA5 antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder