MilliporeSigma
  • Mechanism of circHIPK3-miRNA-124-3p/miRNA-148b-3p-Mediated Inflammatory Responses and Cell Senescence in Candida albicans-Induced Septic Acute Kidney Injury.
Mechanism of circHIPK3-miRNA-124-3p/miRNA-148b-3p-Mediated Inflammatory Responses and Cell Senescence in Candida albicans-Induced Septic Acute Kidney Injury.
Gerontology (2022-05-17)
Jing Han, Wanli Li, Jie Zhang, Yulin Guan, Ying Huang, Xiaoyue Li
ABSTRACT
Sepsis is a life-threatening inflammatory state that can result in septic acute kidney injury (SAKI). Circular RNAs (circRNAs) are implicated in various inflammatory diseases including SAKI. This study investigated the effect of circHIPK3 on inflammatory responses and cell senescence in Candida albicans-induced SAKI. circHIPK3 expression and inflammatory factors in the serum of SAKI patients and healthy volunteers were detected. The murine and cell models of SAKI were established by C. albicans and lipopolysaccharide induction, respectively. The effect of circHIPK3 on SAKI inflammatory responses and cell senescence was measured using ELISA, SA-β-gal staining, CCK-8, RT-qPCR, and Western blot. The binding relationships among circHIPK3, miR-124-3p, or miR-148b-3p and KLF6 or DNMT1/3a were confirmed. The binding of KLF6 and NLRP3 was determined, and the methylation level of the Klotho promoter was detected. Functional rescue experiments were performed to verify the effect of miR-124-3p or miR-148b-3p on SAKI. circHIPK3 was highly expressed in SAKI. circHIPK3 silencing alleviated kidney injury in SAKI mice and enhanced SAKI cell viability by alleviating inflammatory responses and cell senescence. Mechanically, circHIPK3 upregulated KLF6 expression by competitively binding to miR-124-3p, thereby promoting the binding of KLF6 and NLRP3, activating NLRP3/caspase-1-mediated pyroptosis, and eventually aggravating SAKI inflammatory responses. circHIPK3 upregulated DNMT1/3a expression by competitively binding to miR-148b-3p, thus elevating the methylation level of Klotho promoter and accelerating SAKI cell senescence. Downregulation of miR-124-3p or miR-148b-3p attenuated the protective effect of circHIPK3 silencing on SAKI. circHIPK3 aggravated SAKI inflammatory responses via miR-124-3p/KLF6 and accelerated SAKI cell senescence via miR-148b-3p/DNMT1/3a.
Sigma-Aldrich
Anti-KLF6, affinity isolated antibody
Sigma-Aldrich
Senescence Cells Histochemical Staining Kit, sufficient for 100 tests