MilliporeSigma
  • Primate hemorrhagic fever-causing arteriviruses are poised for spillover to humans.
Primate hemorrhagic fever-causing arteriviruses are poised for spillover to humans.
Cell (2022-10-02)
Cody J Warren, Shuiqing Yu, Douglas K Peters, Arturo Barbachano-Guerrero, Qing Yang, Bridget L Burris, Gabriella Worwa, I-Chueh Huang, Gregory K Wilkerson, Tony L Goldberg, Jens H Kuhn, Sara L Sawyer
ABSTRACT
Simian arteriviruses are endemic in some African primates and can cause fatal hemorrhagic fevers when they cross into primate hosts of new species. We find that CD163 acts as an intracellular receptor for simian hemorrhagic fever virus (SHFV; a simian arterivirus), a rare mode of virus entry that is shared with other hemorrhagic fever-causing viruses (e.g., Ebola and Lassa viruses). Further, SHFV enters and replicates in human monocytes, indicating full functionality of all of the human cellular proteins required for viral replication. Thus, simian arteriviruses in nature may not require major adaptations to the human host. Given that at least three distinct simian arteriviruses have caused fatal infections in captive macaques after host-switching, and that humans are immunologically naive to this family of viruses, development of serology tests for human surveillance should be a priority.
Product Description
Millipore
Benzonase® Nuclease, ≥250 units/μL, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous glycerol solution
Sigma-Aldrich
Interleukin-4 human, IL-4, potency:, recombinant, expressed in HEK 293 cells, HumanKine®, suitable for cell culture