MilliporeSigma
  • miR-3059-3p Regulates Glioblastoma Multiforme Radiosensitivity Enhancement through the Homologous Recombination Pathway of DNA Repair.
miR-3059-3p Regulates Glioblastoma Multiforme Radiosensitivity Enhancement through the Homologous Recombination Pathway of DNA Repair.
Journal of oncology (2022-10-04)
Yu-Wen Cheng, Chien-Ju Lin, Shih-Hsun Kuo, Ann-Shung Lieu, Chee-Yin Chai, Hung-Pei Tsai, Aij-Lie Kwan
ABSTRACT
Glioblastoma multiforme (GBM) is one of the most deadly and recalcitrant illnesses of the neurocentral nervous system in humans. MicroRNAs (miRNAs) are a class of noncoding RNAs that play important roles in the regulation of gene expression and biological processes, including radiosensitivity. In this study, we demonstrated the relationship between miR-3059-3p and radiation in GBM. Radioresistant (RR) cells were obtained by exposing GBM8401 cells to 80 Gy radiation in 20 weekly 4 Gy fractions. miR-3059-3p mRNA and DNA replication helicase/nuclease 2 (DNA2) protein expressions were detected using real-time polymerase chain reaction and immunoblotting. Using flow cytometry, colony formation and apoptosis were identified using miR-3059-3p mimic, miR-3059-3p inhibitor, DNA2 siRNA, and DNA2 plasmid. Immunoblotting was used to detect DNA repair proteins. Low levels of miR-3059-3p and high levels of DNA2 were observed in RR cells. Colony formation and apoptosis assays revealed that miR-3059-3p targeted DNA2 to regulate radioresistance. Immunoblotting revealed that miR-3059-3p regulated the homologous recombination (HR) pathway (Rad51 and Rad52) but not the nonhomologous end joining pathway (ku70 and ku80). Downregulation of DNA2 via miR-3059-3p enhanced the radiosensitivity of GBM cells through the inhibition of the HR pathway.
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®
Sigma-Aldrich
Anti-DNA2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, Peroxidase Conjugated, H+L, 1 mg/mL (after reconstitution), Chemicon®