MilliporeSigma
  • AKT phosphorylation as a predictive biomarker for PI3K/mTOR dual inhibition-induced proteolytic cleavage of mTOR companion proteins in small cell lung cancer.
AKT phosphorylation as a predictive biomarker for PI3K/mTOR dual inhibition-induced proteolytic cleavage of mTOR companion proteins in small cell lung cancer.
Cell & bioscience (2022-08-03)
Ming-Chun Hung, Wan-Ping Wang, Ya-Hui Chi
ABSTRACT
Constitutive activation of PI3K signaling has been well recognized in a subset of small cell lung cancer (SCLC), the cancer type which has the most aggressive clinical course amongst pulmonary tumors. Whereas cancers that acquire a mutation/copy gain in PIK3CA or loss of PTEN have been implicated in enhanced sensitivity to inhibitors targeting the PI3K/AKT/mTOR pathway, the complexities of the pathway and corresponding feedback loops hamper clear predictions as to the response of tumors presenting these genomic features. The correlation between the expression profile of proteins involved in the PI3K/AKT/mTOR signaling and cell viability in response to treatment with small molecule inhibitors targeting isoform-specific PI3Ks, AKT, and mTOR was assessed in 13 SCLC cancer cell lines. Athymic nude mice were used to determine the effect of PI3K/mTOR dual inhibition on the growth of xenograft SCLC tumors in vivo. The activation of caspase signaling and proteolytic cleavages of mTOR companion proteins were assessed using recombinant caspases assays and Western blot analyses. Our results indicate that the sensitivity of these SCLC cell lines to GSK2126458, a dual PI3K/mTOR inhibitor, is positively correlated with the expression levels of phosphorylated AKT (p-AKT) at Thr308 and Ser473. Inhibition of pan-class I PI3Ks or PI3K/mTOR dual inhibition was shown to induce proteolytic cleavage of RICTOR and RPTOR, which were respectively dependent on Caspase-6 and Caspase-3. A combination of a clinically approved PI3Kα-selective inhibitor and an mTORC1 inhibitor was shown to have synergistic effects in inducing the death of SCLC cells with high p-AKT. We observed no clear correlation between PTEN levels and the survival of SCLCs in response to PI3K/mTOR dual inhibition; however, PTEN depletion was shown to increase the susceptibility of low p-AKT SCLC cells to dual PI3K/mTOR inhibitor-induced cell death as well as the proteolytic cleavage of RICTOR. These results suggest the level of p-AKT can be a companion diagnostic biomarker for the treatment of SCLC involving the combinational use of clinically approved isoform-specific PI3K and mTOR inhibitors.
Sigma-Aldrich
Anti-Akt1/PKBα Antibody, clone AW24 | 04-796, clone AW24, from rabbit
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture