MilliporeSigma
New putative phenol oxidase in ascidian blood cells.
Scientific reports (2022-08-23)
M A Daugavet, M I Dobrynina, T G Shaposhnikova, A I Solovyeva, A G Mittenberg, S V Shabelnikov, I Yu Babkina, A V Grinchenko, D V Ilyaskina, O I Podgornaya
ABSTRACT
The phenol oxidase system is ancient and ubiquitously distributed in all living organisms. In various groups it serves for the biosynthesis of pigments and neurotransmitters (dopamine), defence reactions and tissue hardening. Ascidians belong to subphylum Tunicata, which is considered the closest living relative to Vertebrates. Two phenol oxidases previously described for ascidians are vertebrate-like and arthropod-like phenol oxidases. In our present study, we described a new ascidian protein, Tuphoxin, with putative phenol oxidase function, which bears no sequence similarity with two enzymes described previously. The closest related proteins to Tuphoxin are mollusc haemocyanins. Unlike haemocyanins, which are oxygen transporting plasma proteins, Tuphoxin is synthesised in ascidian blood cells and secreted in the extracellular matrix of the tunic-ascidian outer coverings. Single mature transcript coding for this phenol oxidase can give several protein products of different sizes. Thus limited proteolysis of the initial protein is suggested. A unique feature of Tuphoxins and their homologues among Tunicata is the presence of thrombospondin first type repeats (TSP1) domain in their sequence which is supposed to provide interaction with extracellular matrix. The finding of TSP1 in the structure of phenol oxidases is new and we consider this to be an innovation of Tunicata evolutionary lineage.
Sigma-Aldrich
Anti-Guinea Pig IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Guinea Pig IgG (whole molecule)−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution