MilliporeSigma
  • Promoting Formation of Oxygen Vacancies in Two-Dimensional Cobalt-Doped Ceria Nanosheets for Efficient Hydrogen Evolution.
Promoting Formation of Oxygen Vacancies in Two-Dimensional Cobalt-Doped Ceria Nanosheets for Efficient Hydrogen Evolution.
Journal of the American Chemical Society (2020-03-24)
Shuaihu Jiang, Ruya Zhang, Hongxian Liu, Yuan Rao, Yanan Yu, Shan Chen, Qin Yue, Yanning Zhang, Yijin Kang
ABSTRACT
As an alternative for depleting fossil fuel energy, hydrogen economy desires low-cost and efficient hydrogen production from water splitting. In order to explore a cheap, abundant, active, and durable catalyst for the electrocatalytic hydrogen evolution reaction (HER), two-dimensional (2D) ceria nanosheets are produced through a thermal decomposition exfoliation method from CeCO3OH with a layer-stacked structure. The additional cobalt dopant promotes formation of oxygen vacancies in ceria nanosheets and, in turn, optimizes hydrogen binding/water dissociation and increases the active sites. As a result, the 2D Co-doped CeO2 nanosheets exhibit an excellent catalytic performance in alkaline HER such that the overpotential is as low as 132 and 215 mV to deliver a high current density of 100 and 500 mA cm-2, respectively, outperforming Pt. Such 2D Co-doped CeO2 nanosheets are also durable HER electrocatalysts, as the activity loss during an extended period of operation is nearly negligible.
Sigma-Aldrich
Cerium(III) nitrate hexahydrate, 99.99% trace metals basis