MilliporeSigma
  • The impact of neuronal Notch-1/JNK pathway on intracerebral hemorrhage-induced neuronal injury of rat model.
The impact of neuronal Notch-1/JNK pathway on intracerebral hemorrhage-induced neuronal injury of rat model.
Oncotarget (2016-09-23)
Maohua Chen, Jun Sun, Chuan Lu, Xiandong Chen, Huajun Ba, Qun Lin, Jianyong Cai, Junxia Dai
ABSTRACT
Notch signaling is a highly conserved pathway that regulates cell fate decisions during embryonic development. Notch activation endangers neurons by modulating NF-κB and HIF-1α pathways, however, the role of Notch signaling in activating JNK/c-Jun following intracerebral hemorrhage (ICH) has not been investigated. In this study, we used rat ICH models and thrombin-induced cell models to investigate the potential role of Notch-1/JNK signals. Our findings revealed that Notch-1 and JNK increased in hematoma-surrounding neurons tissues following ICH during ischemic conditions (all p<0.05). Besides, the expression of active caspase-3 protein was also up-regulated after ICH. According to in-vitro assays, the expression of Notch-1, p-JNK, and active caspase-3 were all up-regulated in cell viability-decreasing ICH cell models (all p<0.05). However, blocking of either Notch-1 or JNK suppressed the phosphorylation of JNK and the expression of active caspase-3, and cell viability was obviously ameliorated. In conclusion, this work suggested Notch-1 activates JNK pathway to induce the active caspase-3, leading to neuronal injury when intracerebral hemorrhage or ischemia occurred. Thus the Notch-1/JNK signal pathway has an important role in ICH process, and may be a therapeutic target to prevent brain injury.
Sigma-Aldrich
AS602801, ≥98% (HPLC)