MilliporeSigma
  • Hyaluronan supports the limbal stem cell phenotype during ex vivo culture.
Hyaluronan supports the limbal stem cell phenotype during ex vivo culture.
Stem cell research & therapy (2022-07-31)
Sudan Puri, Isabel Y Moreno, Mingxia Sun, Sudhir Verma, Xiao Lin, Tarsis F Gesteira, Vivien J Coulson-Thomas
ABSTRACT
Hyaluronan (HA) has previously been identified as an integral component of the limbal stem cell niche in vivo. In this study, we investigated whether a similar HA matrix is also expressed in vitro providing a niche supporting limbal epithelial stem cells (LESCs) during ex vivo expansion. We also investigated whether providing exogenous HA in vitro is beneficial to LESCs during ex vivo expansion. Human LESCs (hLESCs) were isolated from donor corneas and a mouse corneal epithelial progenitor cell line (TKE2) was obtained. The HA matrix was identified surrounding LESCs in vitro using immunocytochemistry, flow cytometry and red blood exclusion assay. Thereafter, LESCs were maintained on HA coated dishes or in the presence of HA supplemented in the media, and viability, proliferation, cell size, colony formation capabilities and expression of putative stem cell markers were compared with cells maintained on commonly used coated dishes. hLESCs and TKE2 cells express an HA-rich matrix in vitro, and this matrix is essential for maintaining LESCs. Further supplying exogenous HA, as a substrate and supplemented to the media, increases LESC proliferation, colony formation capabilities and the expression levels of putative limbal stem cell markers. Our data show that both exogenous and endogenous HA help to maintain the LESC phenotype. Exogenous HA provides improved culture conditions for LESC during ex vivo expansion. Thus, HA forms a favorable microenvironment for LESCs during ex vivo expansion and, therefore, could be considered as an easy and cost-effective substrate and/or supplement for culturing LESCs in the clinic.
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Plasminogen from bovine plasma, lyophilized powder, ≥2.0 units/mg protein
Sigma-Aldrich
Hyaluronidase from Streptomyces hyalurolyticus