MilliporeSigma
  • The conserved C-terminal residues of FAM83H are required for the recruitment of casein kinase 1 to the keratin cytoskeleton.
The conserved C-terminal residues of FAM83H are required for the recruitment of casein kinase 1 to the keratin cytoskeleton.
Scientific reports (2022-07-14)
Takahisa Kuga, Naoki Inoue, Kensuke Sometani, Shino Murataka, Minami Saraya, Rina Sugita, Toshinari Mikami, Yasunori Takeda, Masanari Taniguchi, Kentaro Nishida, Nobuyuki Yamagishi
ABSTRACT
The casein kinase 1 (CK1) family of serine/threonine protein kinases is involved in diverse cellular events at discrete subcellular compartments. FAM83H acts as a scaffold protein that recruits CK1 to the keratin cytoskeleton or to the nuclear speckles, which are storage sites for splicing factors. We determined the amino acid region of FAM83H required for recruiting CK1 to the keratin cytoskeleton. The subcellular localization of mutant FAM83H proteins with deletions of amino acid residues at different positions was evaluated via immunofluorescence. FAM83H mutants with deleted C-terminal residues 1134-1139, which are conserved among vertebrates, lost the ability to localize and recruit CK1 to the keratin cytoskeleton, suggesting that these residues are required for recruiting CK1 to the keratin cytoskeleton. The deletion of these residues (1134-1139) translocated FAM83H and CK1 to the nuclear speckles. Amino acid residues 1 to 603 of FAM83H were determined to contain the region responsible for the recruitment of CK1 to the nuclear speckles. Our results indicated that FAM83H recruits CK1 preferentially to the keratin cytoskeleton and alternatively to the nuclear speckles.
Sigma-Aldrich
Anti-SON antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid
Sigma-Aldrich
Anti-FAM83H antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2