MilliporeSigma
  • Pathogen-selective killing by guanylate-binding proteins as a molecular mechanism leading to inflammasome signaling.
Pathogen-selective killing by guanylate-binding proteins as a molecular mechanism leading to inflammasome signaling.
Nature communications (2022-07-30)
Shouya Feng, Daniel Enosi Tuipulotu, Abhimanu Pandey, Weidong Jing, Cheng Shen, Chinh Ngo, Melkamu B Tessema, Fei-Ju Li, Daniel Fox, Anukriti Mathur, Anyang Zhao, Runli Wang, Klaus Pfeffer, Daniel Degrandi, Masahiro Yamamoto, Patrick C Reading, Gaetan Burgio, Si Ming Man
ABSTRACT
Inflammasomes are cytosolic signaling complexes capable of sensing microbial ligands to trigger inflammation and cell death responses. Here, we show that guanylate-binding proteins (GBPs) mediate pathogen-selective inflammasome activation. We show that mouse GBP1 and GBP3 are specifically required for inflammasome activation during infection with the cytosolic bacterium Francisella novicida. We show that the selectivity of mouse GBP1 and GBP3 derives from a region within the N-terminal domain containing charged and hydrophobic amino acids, which binds to and facilitates direct killing of F. novicida and Neisseria meningitidis, but not other bacteria or mammalian cells. This pathogen-selective recognition by this region of mouse GBP1 and GBP3 leads to pathogen membrane rupture and release of intracellular content for inflammasome sensing. Our results imply that GBPs discriminate between pathogens, confer activation of innate immunity, and provide a host-inspired roadmap for the design of synthetic antimicrobial peptides that may be of use against emerging and re-emerging pathogens.
Product Description
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel - Immunology Multiplex Assay, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.
Millipore
MILLIPLEX® Mouse Cytokine/Chemokine Magnetic Bead Panel II, MECY2MAG-73K, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in mouse serum, plasma and cell culture samples.