MilliporeSigma
  • Discovery of Cathepsin S Inhibitor LY3000328 for the Treatment of Abdominal Aortic Aneurysm.
Discovery of Cathepsin S Inhibitor LY3000328 for the Treatment of Abdominal Aortic Aneurysm.
ACS medicinal chemistry letters (2014-10-15)
Prabhakar K Jadhav, Matthew A Schiffler, Kostas Gavardinas, Euibong J Kim, Donald P Matthews, Michael A Staszak, D Scott Coffey, Bruce W Shaw, Kenneth C Cassidy, Richard A Brier, Yuke Zhang, Robert M Christie, William F Matter, Keyun Qing, Jim D Durbin, Yong Wang, Gary G Deng
ABSTRACT
Cathepsin S (Cat S) plays an important role in many pathological conditions, including abdominal aortic aneurysm (AAA). Inhibition of Cat S may provide a new treatment for AAA. To date, several classes of Cat S inhibitors have been reported, many of which form covalent interactions with the active site Cys25. Herein, we report the discovery of a novel series of noncovalent inhibitors of Cat S through a medium-throughput focused cassette screen and the optimization of the resulting hits. Structure-based optimization efforts led to Cat S inhibitors such as 5 and 9 with greatly improved potency and drug disposition properties. This series of compounds binds to the S2 and S3 subsites without interacting with the active site Cys25. On the basis of in vitro potency, selectivity, and efficacy in a CaCl2-induced AAA in vivo model, 5 (LY3000328) was selected for clinical development.
Sigma-Aldrich
LY3000328, ≥95% (HPLC)