MilliporeSigma
  • Determination of tetracycline antibiotics in beef and pork tissues using ion-paired liquid chromatography.
Determination of tetracycline antibiotics in beef and pork tissues using ion-paired liquid chromatography.
Journal of agricultural and food chemistry (2000-07-11)
W A Moats
ABSTRACT
A simplified procedure was developed for determination of tetracycline antibiotics in tissues which improved stability of these compounds in sample extracts and eliminated the need for troublesome cleanup procedures. Tissues were homogenized in water. Acetonitrile (16 mL) and then 1 mL of 0.1 M H(3)PO(4) were added to 4 mL of homogenate and the clear supernatant was filtered. The filtrate was mixed with hexane and dichloromethane and the resulting water layer was collected, evaporated to 1-2 mL, and filtered into autosampler vials. Ion-pairing liquid chromatography was used to separate tetracyclines from interferences in sample extracts, eliminating the need for further cleanup. Analysis was isocratic using a Phenomonex Prodigy ODS(3) column with a mobile phase of 4 mM oxalic acid, 4 mM sodium oxalate, 10 mM sodium decanesulfonate-acetonitrile (70 + 30 for oxytetracycline and tetracycline; 66 + 34 for chlortetracycline). Recoveries were generally in the 90-100% range with limits of quantitation of 0. 05-0.1 ppm. The procedure was evaluated with beef and pork muscle, liver, and kidney.
Sigma-Aldrich
Sodium 1-decanesulfonate, ~98%