MilliporeSigma
  • Minocycline reverses developmental arsenic exposure-induced microglia activation and functional alteration in BALB/c mice.
Minocycline reverses developmental arsenic exposure-induced microglia activation and functional alteration in BALB/c mice.
Environmental toxicology and pharmacology (2022-03-31)
Jamal Ahmad Ansari, Shubhendra K Mishra, Rajib K Dey, Opalina Roy, Shaivya Kushwaha, Vikas Singh, Satyakam Patnaik, Debabrata Ghosh
ABSTRACT
Arsenic activates microglia and exerts bystander effects on neuron. The present study is focused to test whether minocycline, a second generation antibiotic, can reverse the effect of developmental arsenic exposure on microglial activation and function. Pregnant Balb/c dams were gavaged with sodium arsenite (0.38 mg/kg bd wt) from gestational day 5 (GD5) till post natal day 21 (PND21) and then one group of pups continued till PND59 with arsenic gavage. Minocycline (33 mg/kg bd wt) was administered intraperitoneally two weeks till sacrifice, every alternate day. Mice were sacrificed on PND22 and PND60 and used for various assays. Primary microglial were isolated (ex vivo microglia) from experimental animals and used to measure reactive oxygen species (ROS), nitric oxide (NO), cytokine production and phagocytosis. The whole brain lysate was used for western blot analysis of microglial marker CD68 and synaptic marker, post synaptic density protein 95 (PSD95). For real-time PCR analysis of triggering receptor expressed on myeloid cells 2 (TREM2) and PSD95, RNA isolated from whole brain was used. The study reveals that minocycline administration reversed arsenic-induced increased expression of CD68, ROS, NO, cytokine production, phagocytosis and TREM2 expression. Arsenic-induced reduced expression of PSD95 protein was reversed by minocycline, although the mRNA of PSD95 was unaltered among different groups. Finally, we have checked the learning and memory response of the experimental animals using Y-maze test to correlate the arsenic-induced altered level of synaptic protein. Taken together, the present study finds minocycline to reduce arsenic-induced microglial activation and function which in turn reverses the arsenic-induced impaired learning and memory response.
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Pricing and availability is not currently available.