MilliporeSigma
  • Cellulose nanofibrils/polyvinyl acetate nanocomposite adhesives with improved mechanical properties.
Cellulose nanofibrils/polyvinyl acetate nanocomposite adhesives with improved mechanical properties.
Carbohydrate polymers (2016-11-16)
Ons Chaabouni, Sami Boufi
ABSTRACT
Cellulose nanofibrils (CNFs) are nanoscale cellulose produced from renewable resources with strong reinforcing potential when included in a polymer matrix. In this work, the effect of the addition of CNFs on the properties of waterborne polyvinylacetate (PVA) adhesive was investigated. Adhesive formulations with different contents in CNFs from 1 to 10wt.% was prepared by simple mixing of PVA dispersion with CNFs suspension in water. The viscosity of the adhesive increased with the addition of CNFs, namely over a content of 5%. Shear strength of wood joints at dry as well as wet conditions was improved by CNFs inclusion. Adding 10% of CNF to the PVA adhesive increased the shear strength by about 2 folds compared to the neat adhesive. This strengthening effect was explained by the strong reinforcing impact brought by CNFs inclusion in the PVA matrix. The addition of CNFs also improved the water resistance of PVA adhesive and strongly enhanced the mechanical performance in wet conditions, namely over 7% of CNF content.
Sigma-Aldrich
Poly(vinyl acetate), average Mw ~100,000 by GPC, beads