MilliporeSigma
  • Planar yeast estrogen screen with resorufin-β-d-galactopyranoside as substrate.
Planar yeast estrogen screen with resorufin-β-d-galactopyranoside as substrate.
Journal of chromatography. A (2017-04-01)
Dinah Schick, Wolfgang Schwack
ABSTRACT
For the planar yeast estrogen screen (pYES), 4-methylumbelliferyl-β-d-galactopyranoside was generally employed as substrate, delivering blue fluorescing 4-methylumbelliferone after enzymatic cleavage by the YES reporter β-d-galactosidase as the positive signal for the presence of estrogen active compounds (EAC). As environmental samples like waste water also contain blue fluorescent components, it is difficult to differentiate them from pYES signals. Therefore, resorufin-β-d-galactopyranoside (RGP), providing the orange fluorescing resorufin after enzymatic cleavage, was introduced as pYES substrate to determine EAC. With 17β-estradiol (E2) and 17α-ethinylestradiol (EE2), mean limits of detection and quantitation of 3.5 and 6.5pg/zone, respectively, were determined. Obtained recoveries for both E2 and EE2 from spiked water samples in a concentration range of 2-20ng/L were close to 100%. The application of the RGP-pYES on waste water influent and effluent samples showed the clear detection of EAC without interferences. Estrone (E1), Estriol, E2, and an unknown EAC were found in the influent sample (E2 with a mean of 16.9 ng/L and a precision of 11% RSD; n=4), while another unknown EAC was observed in the effluent sample. In addition, the presence of conjugated EAC in the influent was demonstrated by hydrolysis with β-glucuronidase, when the signals of E1 and the unknown increased by about 25% and 100%, respectively.
Sigma-Aldrich
4-Methylumbelliferyl β-D-galactopyranoside, ≥99% (TLC)