MilliporeSigma
  • Association of CD2AP neuronal deposits with Braak neurofibrillary stage in Alzheimer's disease.
Association of CD2AP neuronal deposits with Braak neurofibrillary stage in Alzheimer's disease.
Brain pathology (Zurich, Switzerland) (2021-09-14)
Jessica Camacho, Alberto Rábano, Paula Marazuela, Anna Bonaterra-Pastra, Garazi Serna, Teresa Moliné, Santiago Ramón Y Cajal, Elena Martínez-Sáez, Mar Hernández-Guillamon
ABSTRACT
Genome-wide association studies have described several genes as genetic susceptibility loci for Alzheimer's disease (AD). Among them, CD2AP encodes CD2-associated protein, a scaffold protein implicated in dynamic actin remodeling and membrane trafficking during endocytosis and cytokinesis. Although a clear link between CD2AP defects and glomerular pathology has been described, little is known about the function of CD2AP in the brain. The aim of this study was to analyze the distribution of CD2AP in the AD brain and its potential associations with tau aggregation and β-amyloid (Aβ) deposition. First, we performed immunohistochemical analysis of CD2AP expression in brain tissue from AD patients and controls (N = 60). Our results showed granular CD2AP immunoreactivity in the human brain endothelium in all samples. In AD cases, no CD2AP was found to be associated with Aβ deposits in vessels or parenchymal plaques. CD2AP neuronal inclusions similar to neurofibrillary tangles (NFT) and neuropil thread-like deposits were found only in AD samples. Moreover, immunofluorescence analysis revealed that CD2AP colocalized with pTau. Regarding CD2AP neuronal distribution, a hierarchical progression from the entorhinal to the temporal and occipital cortex was detected. We found that CD2AP immunodetection in neurons was strongly and positively associated with Braak neurofibrillary stage, independent of age and other pathological hallmarks. To further investigate the association between pTau and CD2AP, we included samples from cases of primary tauopathies (corticobasal degeneration [CBD], progressive supranuclear palsy [PSP], and Pick's disease [PiD]) in our study. Among these cases, CD2AP positivity was only found in PiD samples as neurofibrillary tangle-like and Pick body-like deposits, whereas no neuronal CD2AP deposits were detected in PSP or CBD samples, which suggested an association of CD2AP neuronal expression with 3R-Tau-diseases. In conclusion, our findings open a new road to investigate the complex cellular mechanism underlying the tangle conformation and tau pathology in the brain.
Sigma-Aldrich
Anti-Tau (4-repeat isoform RD4) Antibody, clone 1E1/A6, culture supernatant, clone 1E1/A6, Upstate®
Sigma-Aldrich
Anti-CD2AP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Tau (3-repeat isoform RD3) Antibody, clone 8E6/C11, culture supernatant, clone 8E6/C11, Upstate®