MilliporeSigma
  • Ultraviolet-assisted synthesis of polyacrylamide-grafted chitosan nanoparticles and flocculation performance.
Ultraviolet-assisted synthesis of polyacrylamide-grafted chitosan nanoparticles and flocculation performance.
Carbohydrate polymers (2016-07-31)
Jiangya Ma, Kun Fu, Jun Shi, Yongjun Sun, Xinxi Zhang, Lei Ding
ABSTRACT
In the present work, a new flocculant, polyacrylamide-grafted chitosan nanoparticles (NCS-g-PAM), was synthesized by the copolymerization of acrylamide (AM) and chitosan nanoparticle (NCS) under ultraviolet irradiation using 2-hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone as photo-initiator. The NCS was prepared by the ionic gelation between chitosan and sodium tripolyphosphate. The structure and morphology of NCS-g-PAM were characterized by Fourier Transform Infraredspectroscopy (FT-IR), X-ray diffraction, (1)H-nuclear magnetic resonance spectrometry, scanning electron microscopy, and thermogravimetric analysis. The factors affecting the intrinsic viscosity and the yield of copolymer were studied, which showed that the optimum conditions for the synthesis of NCS-g-PAM were mAM:mNCS=8:1, 0.15g of initiator dosage, mCS:mTPP=4.5:1, 1min of ultrasonication time, 4h of illumination time, and 30min of stirring time. The NCS-g-PAM was found to be more effective than NC-g-PAM in the flocculation of both kaolin suspension and Cu(2+) simulated wastewater. With 5mg/L of polyaluminium chloride (PAC) coordinated and 1mg/L of NCS-g-PAM it was confirmed to be appropriate for flocculating kaolin suspension.
Sigma-Aldrich
2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone, 98%