MilliporeSigma
  • hnRNP Q and hnRNP A1 Regulate the Translation of Cofilin in Response to Transient Oxygen-Glucose Deprivation in Hippocampal Neurons.
hnRNP Q and hnRNP A1 Regulate the Translation of Cofilin in Response to Transient Oxygen-Glucose Deprivation in Hippocampal Neurons.
Cells (2021-12-25)
Sung Wook Kim, In Kyung Hong, Mingee Kim, Yun Seon Song, Kyong-Tai Kim
ABSTRACT
Protein aggregates of cofilin and actin have been found in neurons under oxygen-glucose deprivation. However, the regulatory mechanism behind the expression of Cfl1 during oxygen-glucose deprivation remains unclear. Here, we found that heterogeneous nuclear ribonucleoproteins (hnRNP) Q and hnRNP A1 regulate the translation of Cfl1 mRNA, and formation of cofilin-actin aggregates. The interaction between hnRNP A1 and Cfl1 mRNA was interrupted by hnRNP Q under normal conditions, while the changes in the expression and localization of hnRNP Q and hnRNP A1 increased such interaction, as did the translation of Cfl1 mRNA under oxygen-glucose deprived conditions. These findings reveal a new translational regulatory mechanism of Cfl1 mRNA in hippocampal neurons under oxygen-glucose deprivation.
Sigma-Aldrich
Monoclonal Anti-hnRNP-Q antibody produced in mouse, clone 18E4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%