MilliporeSigma
  • Discrete limbal epithelial stem cell populations mediate corneal homeostasis and wound healing.
Discrete limbal epithelial stem cell populations mediate corneal homeostasis and wound healing.
Cell stem cell (2021-05-14)
Anna Altshuler, Aya Amitai-Lange, Noam Tarazi, Sunanda Dey, Lior Strinkovsky, Shira Hadad-Porat, Swarnabh Bhattacharya, Waseem Nasser, Jusuf Imeri, Gil Ben-David, Ghada Abboud-Jarrous, Beatrice Tiosano, Eran Berkowitz, Nathan Karin, Yonatan Savir, Ruby Shalom-Feuerstein
ABSTRACT
The accessibility and transparency of the cornea permit robust stem cell labeling and in vivo cell fate mapping. Limbal epithelial stem cells (LSCs) that renew the cornea are traditionally viewed as rare, slow-cycling cells that follow deterministic rules dictating their self-renewal or differentiation. Here, we combined single-cell RNA sequencing and advanced quantitative lineage tracing for in-depth analysis of the murine limbal epithelium. These analysis revealed the co-existence of two LSC populations localized in separate and well-defined sub-compartments, termed the "outer" and "inner" limbus. The primitive population of quiescent outer LSCs participates in wound healing and boundary formation, and these cells are regulated by T cells, which serve as a niche. In contrast, the inner peri-corneal limbus hosts active LSCs that maintain corneal epithelial homeostasis. Quantitative analyses suggest that LSC populations are abundant, following stochastic rules and neutral drift dynamics. Together these results demonstrate that discrete LSC populations mediate corneal homeostasis and regeneration.
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Tamoxifen, ≥99%
Sigma-Aldrich
Anti-Cytokeratin 14 Antibody, clone LL002, clone LL002, Chemicon®, from mouse
Sigma-Aldrich
Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, suitable for electrophoresis, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, ≥96% (agarose gel electrophoresis)