MilliporeSigma
  • Potently neutralizing and protective anti-human metapneumovirus antibodies target diverse sites on the fusion glycoprotein.
Potently neutralizing and protective anti-human metapneumovirus antibodies target diverse sites on the fusion glycoprotein.
Immunity (2022-08-10)
C Garrett Rappazzo, Ching-Lin Hsieh, Scott A Rush, Emma S Esterman, Teresa Delgado, James C Geoghegan, Anna Z Wec, Mrunal Sakharkar, Vicente Más, Jason S McLellan, Laura M Walker
ABSTRACT
Human metapneumovirus (hMPV) is a leading cause of acute lower respiratory tract infections in high-risk populations, yet there are no vaccines or anti-viral therapies approved for the prevention or treatment of hMPV-associated disease. Here, we used a high-throughput single-cell technology to interrogate memory B cell responses to the hMPV fusion (F) glycoprotein in young adult and elderly donors. Across all donors, the neutralizing antibody response was primarily directed to epitopes expressed on both pre- and post-fusion F conformations. However, we identified rare, highly potent broadly neutralizing antibodies that recognize pre-fusion-specific epitopes and structurally characterized an antibody that targets a site of vulnerability at the pre-fusion F trimer apex. Additionally, monotherapy with neutralizing antibodies targeting three distinct antigenic sites provided robust protection against lower respiratory tract infection in a small animal model. This study provides promising monoclonal antibody candidates for passive immunoprophylaxis and informs the rational design of hMPV vaccine immunogens.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Human Metapneumovirus Antibody, clone 507, clone 507, Chemicon®, from mouse
Sigma-Aldrich
AEC Staining Kit, liquid
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, HRP conjugate, Chemicon®, from rabbit