MilliporeSigma
RNA G-quadruplex in TMPRSS2 reduces SARS-CoV-2 infection.
Nature communications (2022-03-19)
Geng Liu, Wenya Du, Xiongbo Sang, Qiyu Tong, Ye Wang, Guoqing Chen, Yi Yuan, Lili Jiang, Wei Cheng, Dan Liu, Yan Tian, Xianghui Fu
ABSTRACT
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) infection continues to have devastating consequences worldwide. Recently, great efforts have been made to identify SARS-CoV-2 host factors, but the regulatory mechanisms of these host molecules, as well as the virus per se, remain elusive. Here we report a role of RNA G-quadruplex (RG4) in SARS-CoV-2 infection. Combining bioinformatics, biochemical and biophysical assays, we demonstrate the presence of RG4s in both SARS-CoV-2 genome and host factors. The biological and pathological importance of these RG4s is then exemplified by a canonical 3-quartet RG4 within Tmprss2, which can inhibit Tmprss2 translation and prevent SARS-CoV-2 entry. Intriguingly, G-quadruplex (G4)-specific stabilizers attenuate SARS-CoV-2 infection in pseudovirus cell systems and mouse models. Consistently, the protein level of TMPRSS2 is increased in lungs of COVID-19 patients. Our findings reveal a previously unknown mechanism underlying SARS-CoV-2 infection and suggest RG4 as a potential target for COVID-19 prevention and treatment.
Sigma-Aldrich
TMPyP4, A potent inhibitor of human telomerase (IC₅₀ = 6.5 µM).