MilliporeSigma
  • Discovery of ultrapotent broadly neutralizing antibodies from SARS-CoV-2 elite neutralizers.
Discovery of ultrapotent broadly neutralizing antibodies from SARS-CoV-2 elite neutralizers.
Cell host & microbe (2022-01-02)
Kanika Vanshylla, Chengcheng Fan, Marie Wunsch, Nareshkumar Poopalasingam, Matthijs Meijers, Christoph Kreer, Franziska Kleipass, Denis Ruchnewitz, Meryem S Ercanoglu, Henning Gruell, Friederike Münn, Kai Pohl, Hanna Janicki, Tobias Nolden, Simone Bartl, Saskia C Stein, Max Augustin, Felix Dewald, Lutz Gieselmann, Philipp Schommers, Thomas F Schulz, Leif Erik Sander, Manuel Koch, Marta Łuksza, Michael Lässig, Pamela J Bjorkman, Florian Klein
ABSTRACT
A fraction of COVID-19 convalescent individuals mount a potent antibody response to SARS-CoV-2 with cross-reactivity to SARS-CoV-1. To uncover their humoral response in detail, we performed single B cell analysis from 10 SARS-CoV-2 elite neutralizers. We isolated and analyzed 126 monoclonal antibodies, many of which were sarbecovirus cross-reactive, with some displaying merbecovirus- and embecovirus-reactivity. Several isolated broadly neutralizing antibodies were effective against B.1.1.7, B.1.351, B.1.429, B.1.617, and B.1.617.2 variants and 19 prominent potential escape sites. Furthermore, assembly of 716,806 SARS-CoV-2 sequences predicted emerging escape variants, which were also effectively neutralized. One of these broadly neutralizing potent antibodies, R40-1G8, is a IGHV3-53 RBD-class-1 antibody. Remarkably, cryo-EM analysis revealed that R40-1G8 has a flexible binding mode, targeting both "up" and "down" conformations of the RBD. Given the threat of emerging SARS-CoV-2 variants, we demonstrate that elite neutralizers are a valuable source for isolating ultrapotent antibody candidates to prevent and treat SARS-CoV-2 infection.
Sigma-Aldrich
Papain from papaya latex, buffered aqueous suspension, 2× Crystallized, ≥16 units/mg protein
Protein G Sepharose 4 Fast Flow, Cytiva 17-0618-05, pack of 200 mL