MilliporeSigma
  • Antibiotic synergy interaction against multidrug-resistant Pseudomonas aeruginosa isolated from an abattoir effluent environment.
Antibiotic synergy interaction against multidrug-resistant Pseudomonas aeruginosa isolated from an abattoir effluent environment.
TheScientificWorldJournal (2012-05-26)
Etinosa O Igbinosa, Emmanuel E Odjadjare, Isoken H Igbinosa, Phillips O Orhue, May N O Omoigberale, Napoleon I Amhanre
ABSTRACT
Pseudomonas aeruginosa is an opportunistic pathogen in environmental waters with a high prevalence of multidrug resistance. In this study the synergistic efficacy of synergy antibiotic combinations in multidrug-resistant P. aeruginosa strains isolated from an abattoir effluent was investigated. Water samples were processed using membrane filtration; Pseudomonas was isolated with Pseudomonas Isolation Agar and confirmed using polymerase chain reaction with specie-specific primer. Susceptibility studies and in vitro synergy interaction testing were carried out, employing agar dilution and Etest procedure, respectively. Resistance was noted for clinically relevant antipseudomonal agents tested. Finding from antibiotic synergy interaction studies revealed that cefepime, imipenem, and meropenem combined with amikacin resulted in statistically significant (P < 0.0001) in vitro antibiotics synergy interaction, indicating the possible use of this regimen in treatment of pseudomonal infections.
Sigma-Aldrich
Amikacin Ready Made Solution, 25 mg/mL in water