MilliporeSigma
  • [A case of ureteral endometriosis; the usefulness of MRI for preoperative diagnosis].
[A case of ureteral endometriosis; the usefulness of MRI for preoperative diagnosis].
Hinyokika kiyo. Acta urologica Japonica (1992-02-01)
M Harada, T Kase, M Tajima, Y Sawamura, M Matsushima, M Wakayama, A Kuwajima, M Yokoyama
ABSTRACT
A rare case of ureteral endometriosis is reported. A 46-year-old nulliparous woman was referred to our hospital because of right hydronephrosis which was detected by ultrasonography; Drip infusion pyelography showed right hydroureteronephrosis with the distal obstruction. MRI demonstrated right ovarian endometriosis and resultant ureteral stricture. Based on these findings, we made a preoperative diagnosis of right ureteral endometriosis. Right partial ureterectomy and ureteroureterostomy were performed. Pathological examination of the resected segment showed endometrial tissue around the ureteral lesion. She was administered LH-RH agonist (buserelin); her postoperative course was uneventful. IP showed no abnormalities 6 weeks after the operation. To our knowledge, this case is only the 55th case reported in the literature in Japan.
Sigma-Aldrich
Pinoresinol diglucoside, ≥98% (HPLC)