MilliporeSigma
  • Stroke and Amyloid-β Downregulate TREM-2 and Uch-L1 Expression that Synergistically Promote the Inflammatory Response.
Stroke and Amyloid-β Downregulate TREM-2 and Uch-L1 Expression that Synergistically Promote the Inflammatory Response.
Journal of Alzheimer's disease : JAD (2019-08-28)
Michela Guglielmotto, Ivan Enrico Repetto, Debora Monteleone, Valeria Vasciaveo, Claudio Franchino, Sara Rinaldi, Massimo Tabaton, Elena Tamagno
ABSTRACT
Neuroinflammation is involved in the pathogenesis of Alzheimer's disease, and the transcription factor NF-κB is a player in this event. We found here that the ischemic damage alone or in association with Aβ1-42 activates the NF-κB pathway, induces an increase of BACE1 and a parallel inhibition of Uch-L1 and TREM2, both in vitro and in vivo, in Tg 5XFAD and in human brains of sporadic AD. This mechanism creates a synergistic loop that fosters inflammation. We also demonstrated a significant protection exerted by the restoration of Uch-L1 activity. The rescue of the enzyme is able to abolish the decrease of TREM2 and the parameters of neuroinflammation.
Sigma-Aldrich
Anti-BACE Antibody, CT, Chemicon®, from rabbit
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid