MilliporeSigma
  • Non-canonical initiation factors modulate repeat-associated non-AUG translation.
Non-canonical initiation factors modulate repeat-associated non-AUG translation.
Human molecular genetics (2022-02-28)
Katelyn M Green, Shannon L Miller, Indranil Malik, Peter K Todd
ABSTRACT
Repeat-associated non-AUG (RAN) translation of expanded repeat-mutation mRNA produces toxic peptides in neurons of patients suffering from neurodegenerative diseases. Recent findings indicate that RAN translation in diverse model systems is not inhibited by cellular stressors that impair global translation through phosphorylation of the alpha subunit of eIF2, the essential eukaryotic translation initiation factor that brings the initiator tRNA to the 40S ribosome. Using in vitro, cell-based and Drosophila models, we examined the role of alternative ternary complex factors that may function in place of eIF2, including eIF2A, eIF2D, DENR and MCTS1. Among these factors, DENR knockdown had the greatest inhibitory effect on RAN translation of expanded GGGGCC and CGG repeat reporters and its reduction improved the survival of Drosophila expressing expanded GGGGCC repeats. Taken together, these data support a role for alternative initiation factors in RAN translation and suggest these may serve as novel therapeutic targets in neurodegenerative disease.
Sigma-Aldrich
Monoclonal Anti-DENR antibody produced in mouse, clone 1H3, purified immunoglobulin, buffered aqueous solution