MilliporeSigma
  • Galactose Supplementation in Patients With TMEM165-CDG Rescues the Glycosylation Defects.
Galactose Supplementation in Patients With TMEM165-CDG Rescues the Glycosylation Defects.
The Journal of clinical endocrinology and metabolism (2017-03-23)
Willy Morelle, Sven Potelle, Peter Witters, Sunnie Wong, Leslie Climer, Vladimir Lupashin, Gert Matthijs, Therese Gadomski, Jaak Jaeken, David Cassiman, Eva Morava, François Foulquier
ABSTRACT
TMEM165 deficiency is a severe multisystem disease that manifests with metabolic, endocrine, and skeletal involvement. It leads to one type of congenital disorders of glycosylation (CDG), a rapidly growing group of inherited diseases in which the glycosylation process is altered. Patients have decreased galactosylation by serum glycan analysis. There are >100 CDGs, but only specific types are treatable. Galactose has been shown to be beneficial in other CDG types with abnormal galactosylation. The aim of this study was to characterize the effects of galactose supplementation on Golgi glycosylation in TMEM165-depleted HEK293 cells, as well as in 2 patients with TMEM165-CDG and in their cultured skin fibroblast cells. Glycosylation was assessed by mass spectrometry, western blot analysis, and transferrin isoelectrofocusing. Both unrelated patients with TMEM165-CDG with the same deep intronic homozygous mutation (c.792+182G>A) were allocated to receive d-galactose in a daily dose of 1 g/kg. We analyzed N-linked glycans and glycolipids in knockout TMEM165 HEK293 cells, revealing severe hypogalactosylation and GalNAc transfer defects. Although these defects were completely corrected by the addition of Mn2+, we demonstrated that the observed N-glycosylation defect could also be overcome by galactose supplementation. We then demonstrated that oral galactose supplementation in patients with TMEM165-deficient CDG improved biochemical and clinical parameters, including a substantial increase in the negatively charged transferrin isoforms, and a decrease in hypogalactosylated total N-glycan structures, endocrine function, and coagulation parameters. To our knowledge, this is the first description of abnormal glycosylation of lipids in the TMEM165 defect and the first report of successful dietary treatment in TMEM165 deficiency. We recommend the use of oral d-galactose therapy in TMEM165-CDG.
Sigma-Aldrich
CelLytic MEM Protein Extraction Kit