MilliporeSigma
  • Boosting the peripheral immune response in the skeletal muscles improved motor function in ALS transgenic mice.
Boosting the peripheral immune response in the skeletal muscles improved motor function in ALS transgenic mice.
Molecular therapy : the journal of the American Society of Gene Therapy (2022-04-29)
Maria Chiara Trolese, Carlotta Scarpa, Valentina Melfi, Paola Fabbrizio, Francesca Sironi, Martina Rossi, Caterina Bendotti, Giovanni Nardo
ABSTRACT
Monocyte chemoattractant protein-1 (MCP1) is one of the most powerful pro-inflammatory chemokines. However, its signaling is pivotal in driving injured axon and muscle regeneration. We previously reported that MCP1 is more strongly upregulated in the nervous system of slow-progressing than fast-progressing SOD1G93A mice, the latter showing a poor immune response and eventual massive nerve and muscle degeneration. To assess the MCP1-mediated therapeutic role, we boosted the chemokine along the motor unit of the two SOD1G93A models through a single intramuscular injection of a scAAV9 vector engineered with the Mcp1 gene. We provided direct evidence underlying the pivotal role of the immune response in driving skeletal muscle regeneration and thus the speed of ALS progression. The comparative study performed in fast- and slow-progressing SOD1G93A mice spotlights the nature and temporal activation of the inflammatory response as limiting factors to preserve the periphery and interfere with the disease course. In addition, we recorded a novel pleiotropic role of MCP1 in promoting peripheral axon regeneration and modulating neuroinflammation, ultimately preventing neurodegeneration. Altogether, these observations highlight the immune response as a key determinant for disease variability and proffer a reasonable explanation for the failure of systemic immunomodulatory treatments, suggesting new potential strategies to hamper ALS progression.
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-S100B antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2720, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Neurofilament 200 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Sirt1 antibody, Mouse monoclonal, clone SIR11, purified from hybridoma cell culture