MilliporeSigma
  • Prolonged deprivation of arginine or leucine induces PI3K/Akt-dependent reactivation of mTORC1.
Prolonged deprivation of arginine or leucine induces PI3K/Akt-dependent reactivation of mTORC1.
The Journal of biological chemistry (2022-05-17)
Gwen R Buel, Huy Q Dang, John M Asara, John Blenis, Anders P Mutvei
ABSTRACT
The mechanistic target of rapamycin complex 1 (mTORC1) is a serine/threonine kinase complex that promotes anabolic processes including protein, lipid, and nucleotide synthesis, while suppressing catabolic processes such as macroautophagy. mTORC1 activity is regulated by growth factors and amino acids, which signal through distinct but integrated molecular pathways: growth factors largely signal through the PI3K/Akt-dependent pathway, whereas the availabilities of amino acids leucine and arginine are communicated to mTORC1 by the Rag-GTPase pathway. While it is relatively well described how acute changes in leucine and arginine levels affect mTORC1 signaling, the effects of prolonged amino acid deprivation remain less well understood. Here, we demonstrate that prolonged deprivation of arginine and/or leucine leads to reactivation of mTORC1 activity, which reaches activation levels similar to those observed in nutrient-rich conditions. Surprisingly, we find that this reactivation is independent of the regeneration of amino acids by canonical autophagy or proteasomal degradation but is dependent on PI3K/Akt signaling. Together, our data identify a novel crosstalk between the amino acid and PI3K/Akt signaling pathways upstream of mTORC1. These observations extend our understanding of the role of mTORC1 in growth-related diseases and indicate that dietary intervention by removal of leucine and/or arginine may be an ineffective therapeutic approach.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Puromycin Antibody, clone 12D10, clone 12D10, from mouse