MilliporeSigma
  • The Impact of Some Natural Phenolic Compounds on α-Glucosidase and Sorbitol Dehydrogenase Enzymes, and Anti-leukemia Cancer Potential, Spin Density Distributions, and in silico Studies.
The Impact of Some Natural Phenolic Compounds on α-Glucosidase and Sorbitol Dehydrogenase Enzymes, and Anti-leukemia Cancer Potential, Spin Density Distributions, and in silico Studies.
Journal of oleo science (2022-05-19)
Qiulian He, Peng Ma, Ramin Torshizi
ABSTRACT
In this study, some phenolic compounds including 4-Hexylresorcinol, 5-Pentadecylresorcinol, 5-Tricosylresorcinol, Bilobol, and Urushiol were tested against α-glycosidase enzyme from Saccharomyces cerevisiae and sorbitol dehydrogenase enzymes from sheep liver. These compounds determined good inhibition properties against α-glycosidase and sorbitol dehydrogenase (SDH) enzymes. IC50 values were record in the range of 1.45±0.20-24.532±3.83 μM for α-glycosidase and 6.20±0.96-108.22±18.02 μM for SDH. These inhibitor compounds can be selective drug candidates as anti-diabetic agents, because of they have inhibition properties against both enzymes. In this study, the anti-oxidant activities of the molecules were compared with density functional theory (DFT) calculations. Comparison was made with the experimental enzymes by molecular modeling calculations. In the cellular and molecular part of the recent study, the treated cells with some phenolic compounds were assessed by molecularly targeted therapy (MTT) assay for cytotoxicity and anti-acute lymphoblastic leukemia potentials on Clone 15 HL-60, HL-60, HL-60/MX1, and HL-60/MX2 cell lines. The IC50 of these compounds were µg/mL level against these cell lines.
Sigma-Aldrich
α-Glucosidase from Saccharomyces cerevisiae, recombinant, expressed in proprietary host, lyophilized powder, ≥100 units/mg protein