MilliporeSigma
  • Molecular characterization of myotonic dystrophy fibroblast cell lines for use in small molecule screening.
Molecular characterization of myotonic dystrophy fibroblast cell lines for use in small molecule screening.
iScience (2022-04-29)
Jana R Jenquin, Alana P O'Brien, Kiril Poukalov, Yidan Lu, Jesus A Frias, Hannah K Shorrock, Jared I Richardson, Hormoz Mazdiyasni, Hongfen Yang, Robert W Huigens, David Boykin, Laura P W Ranum, John Douglas Cleary, Eric T Wang, J Andrew Berglund
ABSTRACT
Myotonic dystrophy type 1 (DM1) and type 2 (DM2) are common forms of adult onset muscular dystrophy. Pathogenesis in both diseases is largely driven by production of toxic-expanded repeat RNAs that sequester MBNL RNA-binding proteins, causing mis-splicing. Given this shared pathogenesis, we hypothesized that diamidines, small molecules that rescue mis-splicing in DM1 models, could also rescue mis-splicing in DM2 models. While several DM1 cell models exist, few are available for DM2 limiting research and therapeutic development. Here, we characterize DM1 and DM2 patient-derived fibroblasts for use in small molecule screens and therapeutic studies. We identify mis-splicing events unique to DM2 fibroblasts and common events shared with DM1 fibroblasts. We show that diamidines can partially rescue molecular phenotypes in both DM1 and DM2 fibroblasts. This study demonstrates the potential of fibroblasts as models for DM1 and DM2, which will help meet an important need for well-characterized DM2 cell models.
Sigma-Aldrich
Anti-RBFOX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.