MilliporeSigma
  • Acute Chelation Therapy-Associated Changes in Urine Gadolinium, Self-reported Flare Severity, and Serum Cytokines in Gadolinium Deposition Disease.
Acute Chelation Therapy-Associated Changes in Urine Gadolinium, Self-reported Flare Severity, and Serum Cytokines in Gadolinium Deposition Disease.
Investigative radiology (2021-01-16)
Holden T Maecker, Janet C Siebert, Yael Rosenberg-Hasson, Lorrin M Koran, Miguel Ramalho, Richard C Semelka
ABSTRACT
The aim of this study was to determine the following in patients who have undergone magnetic resonance imaging with gadolinium-based contrast agents (GBCAs) and meet the proposed diagnostic criteria for gadolinium deposition disease (GDD): (1) the effectiveness of chelation therapy (CT) with intravenous Ca-diethylenetriaminepentaacetic acid in removing retained gadolinium (Gd) and factors affecting the amount removed; (2) the frequency of CT-induced Flare, that is, GDD diagnostic symptom worsening, and factors affecting Flare intensity; (3) whether, as reported in a separate cohort, GDD patients' serum cytokine levels differ significantly from those in healthy normal controls and change significantly in response to CT; and (4) whether urine Gd, Flare reaction, and serum cytokine findings in GDD patients are mimicked in non-ill patients described as having gadolinium storage condition (GSC). Twenty-one GDD subjects and 3 GSC subjects underwent CT. Patients provided pre-CT and post-CT 24-hour urine samples for Gd content determination along with pre-CT and 24-hour post-CT serum samples for cytokine analysis. Patients rated potential Flare 24 hours after CT. Pre-CT and post-CT 24-hour urine Gd analyses and Luminex serum cytokine assays were performed blind to patients' GDD and GSC status and all other data except age and sex. Serum cytokine levels in a healthy normal control group of age- and sex-matched subjects drawn from Stanford influenza vaccination studies were measured once, contemporaneously with those of GDD and GSC patients, using the same Luminex assay. Urine Gd amounts increased post-CT by 4 times or more after 87% of the 30 CT sessions. The most important factors appeared to be the time since the last GBCA dose and the cumulative dose received. Urine Gd amounts for GDD and GSC patients fell in the same ranges. All GDD patients, and no GSC patient, reported a Flare 24 hours post-CT. Linear regression found that Flare intensity was significantly predicted by a model including pre- and post-CT Gd amounts and the number of GBCA-enhanced magnetic resonance imaging. Post-CT, multiple cytokines showed strong positive relationships with GDD patients' Flare intensity in multivariable models. The pre-CT serum levels of 12 cytokines were significantly different in GDD patients compared with healthy flu vaccine controls. The small number of GSC patients precluded analogous statistical testing. Post-CT, GDD patients' serum levels of 20 cytokines were significantly decreased, and 2 cytokines significantly increased. These cytokines did not exhibit the same change pattern in the 3 GSC patients. The small number of GSC patients precluded statistical comparisons of GSC to GDD patients' results. In this preliminary study, 24-hour urine Gd content increased markedly and similarly in GDD and GSC patients after Ca-diethylenetriaminepentaacetic acid CT. Post-CT Flare reaction developed only in GDD patients. The current study is the second finding significantly different serum cytokine levels in GDD patients compared with healthy normal controls. These differences and the difference between GDD and GSC patients' Flare and cytokine responses to CT suggest some inflammatory, immunologic, or other physiological differences in patients with GDD. Further research into the treatment and physiological underpinnings of GDD is warranted.
Product Description
Millipore
MILLIPLEX® Human Cytokine/Chemokine Magnetic Bead Panel II - Premixed 23 Plex - Space Saver (Bulk) Packaging, Simultaneously analyze multiple cytokine and chemokine biomarkers with Bead-Based Multiplex Assays using the Luminex technology, in human serum, plasma and cell culture samples.